5-Amino-2-fluoro-4-nitrobenzotrifluoride 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
4-Fluoro-2-nitro-5-(trifluoromethyl)aniline
H64928.03
H64928.06
H64928.03EA
3510
CZK
H64928.03
H64928.06
5-Amino-2-fluoro-4-nitrobenzotrifluoride 98%
5-Amino-2-fluoro-4-nitrobenzotrifluoride
Purity: 98%
CAS No.: 179062-05-6
Molecular Formula: C7H4F4N2O2
Formula Weight: 224.11
Physical Form: Solid
Appearance: Off-white
Beilstein Reference: 14360199
UN No.: UN2811
MDL No.: MFCD03094267
Vzorec:
C₇H₄F₄N₂O₂ Storage Temperature: Ambient |
MDL Number:
MFCD03094267 CAS číslo: 179062-05-6 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...