2-Methyl-5-nitrobenzotrifluoride 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
1-Methyl-4-nitro-2-(trifluoromethyl)benzene
H64924.06
H64924.14
H64924.06EA
1660
CZK
H64924.06
H64924.14
2-Methyl-5-nitrobenzotrifluoride 98%
2-Methyl-5-nitrobenzotrifluoride
Purity: 98%
CAS No.: 89976-12-5
Molecular Formula: C8H6F3NO2
Formula Weight: 205.14
Physical Form: Solid
Sensitivity: Moisture Sensitive
Appearance: Off-white
Beilstein Reference: 2506013
UN No.: UN2811
MDL No.: MFCD01631684
Vzorec:
C₈H₆F₃NO₂ Storage Temperature: Ambient |
MDL Number:
MFCD01631684 CAS číslo: 89976-12-5 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...