Ethyl-2-chloropyrimidine-5-carboxylate 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Ethyl 2-chloropyrimidine-5-carboxylate
H64914.03
H64914.06
H64914.03EA
2350
CZK
H64914.03
H64914.06
Ethyl-2-chloropyrimidine-5-carboxylate 97%
Ethyl-2-chloropyrimidine-5-carboxylate
Purity: 97%
CAS No.: 89793-12-4
Molecular Formula: C7H7ClN2O2
Formula Weight: 186.60
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 52-60°
Boiling Point: 80°/760mm
Beilstein Reference: 142651
MDL No.: MFCD09863164
Vzorec:
C₇H₇ClN₂O₂ MW: 186,60 g/mol Bod varu: 80 °C (760 mmHg) Bod tání: 52…60 °C |
MDL Number:
MFCD09863164 CAS číslo: 89793-12-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...