Tisk…

2,5-Dihydroxyterephthalic acid 97%

Dodavatel: Thermo Fisher Scientific
Varování

H64888.06 H64888.14
H64888.06EA 4160 CZK
H64888.06 H64888.14
2,5-Dihydroxyterephthalic acid 97%
2,5-Dihydroxyterephthalic acid

Purity: 97% CAS No.: 610-92-4 Molecular Formula: C8H6O6 Formula Weight: 198.13 Physical Form: Solid Appearance: Dark grey to green Melting Point: 300° Beilstein Reference: 2696052 MDL No.: MFCD00132933

Vzorec: C₈H₆O₆
MW: 198.13 g/mol
Storage Temperature: Ambient
MDL Number: MFCD00132933
CAS číslo: 610-92-4
EINECS: 210-239-4

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR