4-Bromo-2-nitroanisole 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-Bromo-1-methoxy-2-nitrobenzene
H64877.06
H64877.14
H64877.22
H64877.06EA
1190
CZK
H64877.06
H64877.14
H64877.22
4-Bromo-2-nitroanisole 97%
4-Bromo-2-nitroanisole
Purity: 97%
CAS No.: 33696-00-3
Molecular Formula: C7H6BrNO3
Formula Weight: 232.03
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 85-100°
Beilstein Reference: 2259903
MDL No.: MFCD00055529
Vzorec:
C₇H₆BrNO₃ MW: 232,03 g/mol Bod tání: 85…100 °C |
MDL Number:
MFCD00055529 CAS číslo: 33696-00-3 EINECS: 251-642-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...