Dimethyl-4-chloropyridine-2,6-dicarboxylate 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Dimethyl 4-chloropyridine-2,6-dicarboxylate, Dimethyl 4-Chloro-2,6-pyridinedicarboxylate, 4-Chloro-2,6-pyridinedicarboxylic acid dimethyl ester
H64841.03
H64841.06
H64841.03EA
3020
CZK
H64841.03
H64841.06
Dimethyl-4-chloropyridine-2,6-dicarboxylate 97%
Dimethyl-4-chloropyridine-2,6-dicarboxylate
Purity: 98%
CAS No.: 5371-70-0
Molecular Formula: owderlNO4
Formula Weight: 229.62
Physical Form: Solid
Appearance: Pale yellow
Melting Point: 139-144°
Beilstein Reference: 197075
MDL No.: MFCD02612781
Vzorec:
C₉H₈ClNO₄ MW: 229,616 g/mol Bod tání: 139…144 °C Storage Temperature: Ambient |
MDL Number:
MFCD02612781 CAS číslo: 5371-70-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...