(2-Methyl-3-nitrophenyl)acetic acid 96%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
2-Methyl-3-nitrophenylacetic Acid
H64810.06
H64810.14
H64810.06EA
858
CZK
H64810.06
H64810.14
(2-Methyl-3-nitrophenyl)acetic acid 96%
(2-Methyl-3-nitrophenyl)acetic acid
Purity: 96%
CAS No.: 23876-15-5
Molecular Formula: C9H9NO4
Formula Weight: 195.17
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 132-136°
Beilstein Reference: 2581973
MDL No.: MFCD07782132
Vzorec:
C₉H₉NO₄ MW: 195.17 g/mol Bod tání: 132…136 °C |
MDL Number:
MFCD07782132 CAS číslo: 23876-15-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...