2-Bromo-4-fluoropyridine 97%
Dodavatel: Thermo Scientific
|
Varování
|
H64785.03
H64785.06
H64785.03EA
2850
CZK
H64785.03
H64785.06
2-Bromo-4-fluoropyridine 97%
2-Bromo-4-fluoropyridine
Purity: 97%
CAS No.: 357927-50-5
Molecular Formula: C5H3BrFN
Formula Weight: 175.99
Storage Temperature: 2°C to 8°C
Flash Point: 76°(169°F)
Physical Form: Liquid
Density: 1.699
Refractive Index: 1.5370
Beilstein Reference: 14158224
MDL No.: MFCD09260909
Vzorec:
C₅H₃BrFN MW: 175,99 g/mol Hustota: 1,699 Bod vzplanutí: 76 °C (169 °F) |
MDL Number:
MFCD09260909 CAS číslo: 357927-50-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...