4-(Trifluoromethyl)-1H-imidazole 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
4-(Trifluoromethyl)imidazole
H64711.03
H64711.06
H64711.03EA
1380
CZK
H64711.03
H64711.06
4-(Trifluoromethyl)-1H-imidazole 98%
4-(Trifluoromethyl)-1H-imidazole
Purity: 98%
CAS No.: 33468-69-8
Molecular Formula: C4H3F3N2
Formula Weight: 136.08
Physical Form: Solid
Melting Point: 148-150°
MDL No.: MFCD08458860
Vzorec:
C₄H₃F₃N₂ MW: 136,08 g/mol Bod tání: 148…150 °C Storage Temperature: Ambient |
MDL Number:
MFCD08458860 CAS číslo: 33468-69-8 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...