Methyl-4-chloroacetoacetate 97+%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
Methyl 4-chloro-3-oxobutanoate
Purity: 97+%
CAS No.: 32807-28-6
Molecular Formula: C5H7ClO3
Formula Weight: 150.56
Flash Point: 102°(216°F)
Physical Form: Liquid
Density: 1.305
Refractive Index: 1.4560
Appearance: Colourless
Boiling Point: 85°/4mm
Beilstein Reference: 1759939
UN No.: UN2922
MDL No.: MFCD00000938
Vzorec:
C₅H₇ClO₃ MW: 150,56 g/mol Bod varu: 85 °C (4 mmHg) Hustota: 1,305 Bod vzplanutí: 102 °C (216 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000938 CAS číslo: 32807-28-6 EINECS: 251-230-5 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...