Violuric acid monohydrate ≥97%
Dodavatel: Thermo Scientific
B23931.09
B23931.18
B23931.09EA
1560
CZK
B23931.09
B23931.18
Violuric acid monohydrate ≥97%
Violuric acid monohydrate
Purity: 97%
CAS No.: 26351-19-9
Molecular Formula: C4H3N3O4.H2O
Formula Weight: 175.10 (157.08anhy)
Melting Point: ca 245° dec.
Beilstein Reference: 162342
Merck Reference: 14,10000
MDL No.: MFCD00149401
Vzorec:
C₄H₃N₃O₄·H₂O Bod tání: ∼245 °C (rozkl.) |
MDL Number:
MFCD00149401 CAS číslo: 26351-19-9 Merck Index: 14,10000 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...