Tisk…

2-Amino-4-chloro-3-nitropyridine 95%

Dodavatel: Thermo Fisher Scientific
Varování

H64678.03 H64678.06
H64678.03EA 3230 CZK
H64678.03 H64678.06
2-Amino-4-chloro-3-nitropyridine 95%
2-Amino-4-chloro-3-nitropyridine

Purity: 95% CAS No.: 6980-08-1 Molecular Formula: C5H4ClN3O2 Formula Weight: 173.56 Beilstein Reference: 391939 MDL No.: MFCD04116097

Vzorec: C₅H₄ClN₃O₂
Storage Temperature: Ambient
MDL Number: MFCD04116097
CAS číslo: 6980-08-1

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR