2-Naphthyl boronic acid pinacol ester 97%
Dodavatel: Thermo Fisher Scientific
Synonyms:
2-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)naphthalene, 4,4,5,5-Tetramethyl-2-(2-naphthyl)-1,3,2-dioxaborolane, Naphthalene-2-boronic acid Pinacol ester, 4,4,5,5-Tetramethyl-2-(naphthalen-2-yl)-1,3,2-dioxaborolane
H64671.03
H64671.06
H64671.03EA
1410
CZK
H64671.03
H64671.06
2-Naphthyl boronic acid pinacol ester 97%
2-Naphthyl boronic acid pinacol ester
Purity: 97%
CAS No.: 256652-04-7
Molecular Formula: C16H19BO2
Formula Weight: 254.14
Physical Form: Solid
MDL No.: MFCD05663868
Vzorec:
C₁₆H₁₉BO₂ MW: 254.14 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD05663868 CAS číslo: 256652-04-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...