Tisk…

2-Methylpropanethioamide 95%

Dodavatel: Thermo Scientific
Varování

Synonyms: Thioisobutyramide

H64663.03 H64663.06 H64663.14
H64663.03EA 1490 CZK
H64663.03 H64663.06 H64663.14
2-Methylpropanethioamide 95%
2-Methylpropanethioamide

Purity: 95% CAS No.: 13515-65-6 Molecular Formula: C4H9NS Formula Weight: 103.18 Storage Temperature: -30°C to -10°C Physical Form: Solid Melting Point: 45-48° Beilstein Reference: 1098427 MDL No.: MFCD07369538

Vzorec: C₄H₉NS
MW: 103,183 g/mol
Bod tání: 45…48 °C
MDL Number: MFCD07369538
CAS číslo: 13515-65-6

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR