Tisk…

2-Bromo-1-(o-tolyl)ethanone 98%

Dodavatel: Thermo Fisher Scientific
Varování

H64651.03 H64651.06
H64651.03EA 3570 CZK
H64651.03 H64651.06
2-Bromo-1-(o-tolyl)ethanone 98%
2-Bromo-1-(o-tolyl)ethanone

Purity: 98% CAS No.: 51012-65-8 Molecular Formula: C9H9BrO Formula Weight: 213.07 Boiling Point: 81-83° Beilstein Reference: 508116 MDL No.: MFCD04038955

Vzorec: C₉H₉BrO
Bod varu: 81…83 °C
Storage Temperature: Ambient
MDL Number: MFCD04038955
CAS číslo: 51012-65-8

Objednat

Produktová specifikace

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR