Tisk…

Tetrahydropyran-4-methanol 98%

Dodavatel: Thermo Fisher Scientific
Nebezpečí

Synonyms: 4-(Hydroxymethyl)tetrahydropyran

H64568.03 H64568.06
H64568.03EA 2440 CZK
H64568.03 H64568.06
Tetrahydropyran-4-methanol 98%
Tetrahydropyran-4-methanol

Purity: 98% CAS No.: 14774-37-9 Molecular Formula: C6H12O2 Formula Weight: 116.16 Flash Point: 61°(142°F) Physical Form: Liquid Refractive Index: 1.4600 Boiling Point: 111-115°/15mm Beilstein Reference: 103069 MDL No.: MFCD00457804

Vzorec: C₆H₁₂O₂
MW: 116.16 g/mol
Bod varu: 111…115 °C (15 mmHg)
Bod vzplanutí: 61 °C (142 °F)
Storage Temperature: Ambient
MDL Number: MFCD00457804
CAS číslo: 14774-37-9

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR