Tisk…

5-Bromo-2,3-difluoropyridine 97%

Dodavatel: Thermo Scientific
Varování

H64566.06 H64566.14
H64566.06EA 5600 CZK
H64566.06 H64566.14
5-Bromo-2,3-difluoropyridine 97%
5-Bromo-2,3-difluoropyridine

Purity: 97% CAS No.: 89402-44-8 Molecular Formula: C5H2BrF2N Formula Weight: 193.98 Physical Form: Liquid Appearance: Colourless to pale yellow Beilstein Reference: 13412561 MDL No.: MFCD06659525

Vzorec: C₅H₂BrF₂N
MW: 193,98 g/mol
Storage Temperature: Ambient
MDL Number: MFCD06659525
CAS číslo: 89402-44-8

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR