Tisk…

1-(Tetrahydro-2H-pyran-4-yl)ethanone ≥97%

Dodavatel: Thermo Scientific
Varování

Synonyms: 4-Acetyltetrahydropyran

H64522.MD H64522.03
H64522.MDEA 4160 CZK
H64522.MD H64522.03
1-(Tetrahydro-2H-pyran-4-yl)ethanone ≥97%
1-(Tetrahydro-2H-pyran-4-yl)ethanone

Purity: 97% CAS No.: 137052-08-5 Molecular Formula: C7H12O2 Formula Weight: 128.17 Physical Form: Liquid Density: 1.024 Refractive Index: 1.4530 Appearance: Colourless to pale yellow Boiling Point: 90-94°/15mm Beilstein Reference: 109587 MDL No.: MFCD08704647

Vzorec: C₇H₁₂O₂
MW: 128,17 g/mol
Bod varu: 90…94 °C (15 mmHg)
Hustota: 1,024
Storage Temperature: Ambient
MDL Number: MFCD08704647
CAS číslo: 137052-08-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR