5-Bromo-1,2,3-trichlorobenzene 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H64506.06
H64506.14
H64506.06EA
1220
CZK
H64506.06
H64506.14
5-Bromo-1,2,3-trichlorobenzene 98%
5-Bromo-1,2,3-trichlorobenzene
Purity: 98%
CAS No.: 21928-51-8
Molecular Formula: C6H2BrCl3
Formula Weight: 260.34
Physical Form: Powder
Appearance: Off-white
Beilstein Reference: 2089937
MDL No.: MFCD00155009
Vzorec:
C₆H₂BrCl₃ Storage Temperature: Ambient |
MDL Number:
MFCD00155009 CAS číslo: 21928-51-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...