2,6-Dimethylbenzaldehyde 97%
Dodavatel: Thermo Fisher Scientific
Synonyms:
2-Formyl-m-xylene
H64503.06
H64503.14
H64503.06EA
1820
CZK
H64503.06
H64503.14
2,6-Dimethylbenzaldehyde 97%
2,6-Dimethylbenzaldehyde
Purity: 97%
CAS No.: 1123-56-4
Molecular Formula: C9H10O
Formula Weight: 134.18
Flash Point: 96°(205°F)
Physical Form: Solid
Refractive Index: 1.5480
Sensitivity: Air Sensitive
Appearance: White to yellow
Melting Point: 27-30° lit.
Boiling Point: 230° lit.
Beilstein Reference: 907019
MDL No.: MFCD00128003
Vzorec:
C₉H₁₀O MW: 134.18 g/mol Bod varu: 230 °C (lit.) Bod tání: 27…30 °C (lit.) Bod vzplanutí: 96 °C (205 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00128003 CAS číslo: 1123-56-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...