Tisk…

5-Nitropicolinic acid 97+%

Dodavatel: Thermo Scientific
Varování

Synonyms: 5-Nitro-2-pyridinecarboxylic acid, 5-Nitropyridine-2-carboxylic acid, 5-Nitropicolinic acid

H64499.03 H64499.06
H64499.03EA 1350 CZK
H64499.03 H64499.06
5-Nitropicolinic acid 97+%
5-Nitropicolinic acid

Purity: 97+% CAS No.: 30651-24-2 Molecular Formula: C6H4N2O4 Formula Weight: 168.11 Physical Form: Solid Melting Point: 208-212° Beilstein Reference: 139209 MDL No.: MFCD04114182

Vzorec: C₆H₄N₂O₄
MW: 168,11 g/mol
Bod tání: 208…212 °C
Storage Temperature: Ambient
MDL Number: MFCD04114182
CAS číslo: 30651-24-2

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR