Tisk…

5-(Trifluoromethyl)picolinonitrile 98%

Dodavatel: Thermo Scientific
Varování

Synonyms: 2-Cyano-5-(trifluoromethyl)pyridine

H64486.03 H64486.06
H64486.03EA 1200 CZK
H64486.03 H64486.06
5-(Trifluoromethyl)picolinonitrile 98%
5-(Trifluoromethyl)picolinonitrile

Purity: 98% CAS No.: 95727-86-9 Molecular Formula: C7H3F3N2 Formula Weight: 172.11 Physical Form: Solid Beilstein Reference: 13339826 UN No.: UN3439 MDL No.: MFCD01001119

Vzorec: C₇H₃F₃N₂
MW: 172,11 g/mol
Storage Temperature: Ambient
MDL Number: MFCD01001119
CAS číslo: 95727-86-9
ADR: 6.1,III

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR