1,5-Naphthyridine 97+%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
H64484.MD
H64484.03
H64484.06
H64484.MDEA
2090
CZK
H64484.MD
H64484.03
H64484.06
1,5-Naphthyridine 97+%
1,5-Naphthyridine
Purity: 97+%
CAS No.: 254-79-5
Molecular Formula: C8H6N2
Formula Weight: 130.15
Physical Form: Solid
Density: 1.25
Appearance: Off-white
Melting Point: 69-75°
Beilstein Reference: 109350
MDL No.: MFCD00039727
Vzorec:
C₈H₆N₂ MW: 130.15 g/mol Bod tání: 69…75 °C Hustota: 1.25 Storage Temperature: Ambient |
MDL Number:
MFCD00039727 CAS číslo: 254-79-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...