Tisk…

4-iodobenzene-1,2-diamine 95%

Dodavatel: Thermo Fisher Scientific
Varování

H64469.MD H64469.03 H64469.06
H64469.MDEA 1530 CZK
H64469.MD H64469.03 H64469.06
4-iodobenzene-1,2-diamine 95%
4-iodobenzene-1,2-diamine

Purity: 95% CAS No.: 21304-38-1 Molecular Formula: C6H7IN2 Formula Weight: 234.04 Physical Form: Solid Sensitivity: Light Sensitive Melting Point: 73-77° Beilstein Reference: 2716284 UN No.: UN2811 MDL No.: MFCD08669466

Vzorec: C₆H₇IN₂
Bod tání: 73…77 °C
Storage Temperature: Ambient
MDL Number: MFCD08669466
CAS číslo: 21304-38-1
ADR: 6.1,III

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR