Tisk…

2-Isopropoxy-5-pyridineboronic acid 97%

Dodavatel: Thermo Scientific
Varování

Synonyms: (6-Isopropoxypyridin-3-yl)boronic acid, 2-Isoproxypyridine-5-boronic acid

H64461.MD H64461.06
H64461.MDEA 1620 CZK
H64461.MD H64461.06
2-Isopropoxy-5-pyridineboronic acid 97%
2-Isopropoxy-5-pyridineboronic acid

Purity: 97% CAS No.: 870521-30-5 Molecular Formula: C8H12BNO3 Formula Weight: 181.00 Physical Form: Solid MDL No.: MFCD07368870

Vzorec: C₈H₁₂BNO₃
MW: 181,00 g/mol
Storage Temperature: Ambient
MDL Number: MFCD07368870
CAS číslo: 870521-30-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR