3-Bromocarbazole 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
3-Bromo-9H-carbazole
H64437.14
H64437.06
H64437.14EA
14250
CZK
H64437.14
H64437.06
3-Bromocarbazole 98%
3-Bromocarbazole
Purity: 98%
CAS No.: 1592-95-6
Molecular Formula: C12H8BrN
Formula Weight: 246.11
Physical Form: Solid
Appearance: White
Melting Point: 195-199°
Boiling Point: 160°/6mm
Beilstein Reference: 142195
MDL No.: MFCD00222621
Vzorec:
C₁₂H₈BrN Bod varu: 160 °C (6 mmHg) Bod tání: 195…199 °C Storage Temperature: Ambient |
MDL Number:
MFCD00222621 CAS číslo: 1592-95-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...