2,3-Dimethylbenzaldehyde 97%
Dodavatel: Thermo Scientific
H64425.06
H64425.14
H64425.22
H64425.06EA
1230
CZK
H64425.06
H64425.14
H64425.22
2,3-Dimethylbenzaldehyde 97%
2,3-Dimethylbenzaldehyde
Purity: 97%
CAS No.: 5779-93-1
Molecular Formula: C9H10O
Formula Weight: 134.18
Flash Point: 101°(214°F)
Physical Form: Liquid
Density: 1.029
Refractive Index: 1.5530
Appearance: Colourless to pale yellow
Boiling Point: 86-88°/10mm
Beilstein Reference: 2238677
MDL No.: MFCD00798004
Vzorec:
C₉H₁₀O MW: 134,18 g/mol Bod varu: 86…88 °C (10 mmHg) Hustota: 1,029 Bod vzplanutí: 101 °C (214 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00798004 CAS číslo: 5779-93-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...