2-Chloro-3-nitro-5-(trifluoromethyl)pyridine 95%
Dodavatel: Thermo Scientific
|
Varování
|
H64394.03
H64394.06
H64394.14
H64394.03EA
1170
CZK
H64394.03
H64394.06
H64394.14
2-Chloro-3-nitro-5-(trifluoromethyl)pyridine 95%
2-Chloro-3-nitro-5-(trifluoromethyl)pyridine
Purity: 95%
CAS No.: 72587-15-6
Molecular Formula: C6H2ClF3N2O2
Formula Weight: 226.54
Physical Form: Liquid
Appearance: Colourless to pale yellow
Beilstein Reference: 7586778
MDL No.: MFCD06255171
Vzorec:
C₆H₂ClF₃N₂O₂ MW: 226,54 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD06255171 CAS číslo: 72587-15-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...