2-methylpyrimidine-5-carboxylic acid 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
Synonyms:
2-Methylpyrimidine-5-carboxylic acid
H64375.03
H64375.06
H64375.03EA
3180
CZK
H64375.03
H64375.06
2-methylpyrimidine-5-carboxylic acid 97%
2-methylpyrimidine-5-carboxylic acid
Purity: 97%
CAS No.: 5194-32-1
Molecular Formula: C6H6N2O2
Formula Weight: 138.13
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Melting Point: 203-205°
Beilstein Reference: 508773
MDL No.: MFCD08235210
Vzorec:
C₆H₆N₂O₂ MW: 138,126 g/mol Bod tání: 203…205 °C |
MDL Number:
MFCD08235210 CAS číslo: 5194-32-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...