β-Alanine-tert-butylester hydrochloride 95%
Dodavatel: Thermo Scientific
Synonyms:
beta-Alanine tert-Butyl ester hydrochloride, β-Alanine t-butyl ester hydrochloride, H-β-Ala-OtBu·HCl, H-beta-Ala-OtBu·HCl, H-beta-Ala-OtBu.HCl, H-β-Ala-OtBu.HCl, tert-Butyl 3-aminopropanoate hydrochloride
H64346.06
H64346.14
H64346.22
H64346.06EA
2550
CZK
H64346.06
H64346.14
H64346.22
β-Alanine-tert-butylester hydrochloride 95%
beta-Alanine-tert-butylester hydrochloride
Purity: 95%
CAS No.: 58620-93-2
Molecular Formula: C7H15NO2.HCl
Formula Weight: 181.66
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Melting Point: 167-169°
Beilstein Reference: 6660122
MDL No.: MFCD00058258
Vzorec:
C₇H₁₆ClNO₂ MW: 181,66 g/mol Bod tání: 167…169 °C |
MDL Number:
MFCD00058258 CAS číslo: 58620-93-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...