Tetrahydropyran-4-carboxylic acid 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Tetrahydropyran-4-yl-carboxylic acid
H64340.06
H64340.14
H64340.06EA
1850
CZK
H64340.06
H64340.14
Tetrahydropyran-4-carboxylic acid 98%
Tetrahydropyran-4-carboxylic acid
Purity: 98%
CAS No.: 5337-03-1
Molecular Formula: C6H10O3
Formula Weight: 130.14
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: White to off-white
Melting Point: 87-89°
Beilstein Reference: 112249
MDL No.: MFCD00031016
Vzorec:
C₆H₁₀O₃ MW: 130,14 g/mol Bod tání: 87…89 °C |
MDL Number:
MFCD00031016 CAS číslo: 5337-03-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...