5,6-Dichloropicolinic acid 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
5,6-Dichloropyridine-2-carboxylic acid
H64320.03
H64320.06
H64320.03EA
3710
CZK
H64320.03
H64320.06
5,6-Dichloropicolinic acid 98%
5,6-Dichloropicolinic acid
Purity: 98%
CAS No.: 88912-24-7
Molecular Formula: C6H3Cl2NO2
Formula Weight: 192.00
Physical Form: Solid
Beilstein Reference: 4390497
MDL No.: MFCD13185546
Vzorec:
C₆H₃Cl₂NO₂ MW: 192.00 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD13185546 CAS číslo: 88912-24-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...