Methyl-4-amino-3-bromobenzoate 97%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
Methyl 4-amino-3-bromobenzoate
H64303.03
H64303.06
H64303.03EA
1690
CZK
H64303.03
H64303.06
Methyl-4-amino-3-bromobenzoate 97%
Methyl-4-amino-3-bromobenzoate
Purity: 97%
CAS No.: 106896-49-5
Molecular Formula: C8H8BrNO2
Formula Weight: 230.06
Physical Form: Solid
Melting Point: 105-109°
Beilstein Reference: 4905039
MDL No.: MFCD01861385
Vzorec:
C₈H₈BrNO₂ MW: 230,06 g/mol Bod tání: 105…109 °C Storage Temperature: Ambient |
MDL Number:
MFCD01861385 CAS číslo: 106896-49-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...