3-Amino-2-bromo-6-methylpyridine 98%
Dodavatel: Thermo Fisher Scientific
|
Nebezpečí
|
Synonyms:
2-Bromo-6-methylpyridin-3-amine, 3-Amino-2-bromo-6-picoline, 5-Amino-6-bromo-2-picoline
H64301.03
H64301.06
H64301.03EA
2550
CZK
H64301.03
H64301.06
3-Amino-2-bromo-6-methylpyridine 98%
3-Amino-2-bromo-6-methylpyridine
Purity: 98%
CAS No.: 126325-53-9
Molecular Formula: C6H7BrN2
Formula Weight: 187.04
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Appearance: Off-white
Beilstein Reference: 4243986
UN No.: UN2811
MDL No.: MFCD03095078
Vzorec:
C₆H₇BrN₂ MW: 187.04 |
MDL Number:
MFCD03095078 CAS číslo: 126325-53-9 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...