Tisk…

4-Fluoro-2-methylpyridine 96%

Dodavatel: Thermo Scientific
Varování

H64284.03 H64284.06
H64284.03EA 2710 CZK
H64284.03 H64284.06
4-Fluoro-2-methylpyridine 96%
4-Fluoro-2-methylpyridine

Purity: 96% CAS No.: 766-16-5 Molecular Formula: C6H6FN Formula Weight: 111.12 Storage Temperature: -30°C to -10°C Physical Form: Liquid Refractive Index: 1.4715 Sensitivity: Air Sensitive Appearance: Colourless to pale yellow Boiling Point: 128-132° Beilstein Reference: 107087 MDL No.: MFCD04114145

Vzorec: C₆H₆FN
MW: 111,12 g/mol
Bod varu: 128…132 °C
MDL Number: MFCD04114145
CAS číslo: 766-16-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR