Diethyl(3-pyridyl)borane 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
3-(Diethylboryl)pyridine
H64225.06
H64225.14
H64225.22
H64225.06EA
1830
CZK
H64225.06
H64225.14
H64225.22
Diethyl(3-pyridyl)borane 98%
Diethyl(3-pyridyl)borane
Purity: 98%
CAS No.: 89878-14-8
Molecular Formula: C9H14BN
Formula Weight: 147.03
Physical Form: Solid
Melting Point: 172-175° lit.
Beilstein Reference: 4381494
MDL No.: MFCD00012348
Vzorec:
C₉H₁₄BN MW: 147,03 g/mol Bod tání: 172…175 °C (lit.) Storage Temperature: Ambient |
MDL Number:
MFCD00012348 CAS číslo: 89878-14-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...