Tisk…

3-Carboxyphenylboronic acid pinacol ester 97%

Dodavatel: Thermo Fisher Scientific

Synonyms: 3-Carboxybenzeneboronic acid pinacol ester, 3-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)benzoic acid

H64195.06 H64195.14
H64195.06EA 2180 CZK
H64195.06 H64195.14
3-Carboxyphenylboronic acid pinacol ester 97%
3-Carboxyphenylboronic acid pinacol ester

Purity: 97% CAS No.: 269409-73-6 Molecular Formula: C13H17BO4 Formula Weight: 248.09 Physical Form: Powder Appearance: White Melting Point: 206-210° dec. MDL No.: MFCD03411930

Vzorec: C₁₃H₁₇BO₄
Storage Temperature: Ambient
MDL Number: MFCD03411930
CAS číslo: 269409-73-6

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR