1-Methyl-1,2,3,4-tetrahydroisoquinoline 95%
Dodavatel: Thermo Scientific
|
Varování
|
H64191.03
H64191.06
H64191.03EA
1780
CZK
H64191.03
H64191.06
1-Methyl-1,2,3,4-tetrahydroisoquinoline 95%
1-Methyl-1,2,3,4-tetrahydroisoquinoline
Purity: 95%
CAS No.: 4965-09-7
Molecular Formula: C10H13N
Formula Weight: 147.22
Storage Temperature: -30°C to -10°C
Physical Form: Liquid
Appearance: Pale yellow
Boiling Point: 119°/18mm
Beilstein Reference: 82140
MDL No.: MFCD00798989
Vzorec:
C₁₀H₁₃N MW: 147,22 g/mol Bod varu: 119 °C (18 mmHg) |
MDL Number:
MFCD00798989 CAS číslo: 4965-09-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...