4-(Trifluoromethyl)anisole 98%
Dodavatel: Thermo Scientific
|
Varování
|
H64187.MD
H64187.03
H64187.06
H64187.MDEA
985
CZK
H64187.MD
H64187.03
H64187.06
4-(Trifluoromethyl)anisole 98%
4-(Trifluoromethyl)anisole
Purity: 98%
CAS No.: 402-52-8
Molecular Formula: C8H7F3O
Formula Weight: 176.14
Storage Temperature: 2°C to 8°C
Flash Point: 60°(140°F)
Physical Form: Liquid
Density: 1.245
Refractive Index: 1.4450
Appearance: Colourless to pale yellow
Melting Point: -9°
Boiling Point: 169°/754mm
UN No.: UN1993
MDL No.: MFCD03094456
Vzorec:
C₈H₇F₃O MW: 176,14 g/mol Bod varu: 169 °C (754 mmHg) Bod tání: –9 °C Hustota: 1,245 Bod vzplanutí: 60 °C (140 °F) |
MDL Number:
MFCD03094456 CAS číslo: 402-52-8 ADR: 3,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...