Tisk…

3,5-Dibromo-4-methoxypyridine 98%

Dodavatel: Thermo Scientific
Varování

H64177.03 H64177.06
H64177.03EA 2460 CZK
H64177.03 H64177.06
3,5-Dibromo-4-methoxypyridine 98%
3,5-Dibromo-4-methoxypyridine

Purity: 98% CAS No.: 650140-84-4 Molecular Formula: C6H5Br2NO Formula Weight: 266.92 Physical Form: Solid Appearance: Off-white MDL No.: MFCD13185524

Vzorec: C6H5Br2NO
MW: 266,920 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13185524
CAS číslo: 25813-24-5

Objednat

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR