2,5-Dimethoxy-4-methylbenzaldehyde 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H64140.06
H64140.14
H64140.06EA
6730
CZK
H64140.06
H64140.14
2,5-Dimethoxy-4-methylbenzaldehyde 97%
2,5-Dimethoxy-4-methylbenzaldehyde
Purity: 97%
CAS No.: 4925-88-6
Molecular Formula: C10H12O3
Formula Weight: 180.20
Physical Form: Solid
Appearance: Off-white
Melting Point: 82-86°
Beilstein Reference: 1866592
MDL No.: MFCD02253192
Vzorec:
C₁₀H₁₂O₃ Bod tání: 82…86 °C Storage Temperature: Ambient |
MDL Number:
MFCD02253192 CAS číslo: 4925-88-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...