7-Bromo-1-heptene 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H64127.06
H64127.14
H64127.22
H64127.06EA
2530
CZK
H64127.06
H64127.14
H64127.22
7-Bromo-1-heptene 97%
7-Bromo-1-heptene
Purity: 97%
CAS No.: 4117-09-3
Molecular Formula: C7H13Br
Formula Weight: 177.08
Flash Point: 58°(136°F)
Physical Form: Liquid
Density: 1.162
Refractive Index: 1.4660
Appearance: Colourless to pale yellow
Boiling Point: 27°/1mm
Beilstein Reference: 1740861
UN No.: UN1993
MDL No.: MFCD01631137
Vzorec:
C₇H₁₃Br Bod varu: 27 °C (1 mmHg) Hustota: 1,162 Bod vzplanutí: 58 °C (136 °F) Storage Temperature: Ambient |
MDL Number:
MFCD01631137 CAS číslo: 4117-09-3 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...