4-Chloro-2-picoline 95%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
4-Chloro-2-methylpyridine
H64093.03
H64093.06
H64093.03EA
1150
CZK
H64093.03
H64093.06
4-Chloro-2-picoline 95%
4-Chloro-2-picoline
Purity: 95%
CAS No.: 3678-63-5
Molecular Formula: C6H6ClN
Formula Weight: 127.57
Physical Form: Liquid
Density: 1.15
Refractive Index: 1.5205
Appearance: Colourless to pale yellow
Boiling Point: 54-56°/15mm lit.
Beilstein Reference: 107196
MDL No.: MFCD00038833
Vzorec:
C₆H₆ClN MW: 127.57 g/mol Bod varu: 54…56 °C (15 mmHg) (lit.) Hustota: 1.15 Storage Temperature: Ambient |
MDL Number:
MFCD00038833 CAS číslo: 3678-63-5 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...