2,5-Dichloropyrimidine 95%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H64066.06
H64066.14
H64066.22
H64066.06EA
1600
CZK
H64066.06
H64066.14
H64066.22
2,5-Dichloropyrimidine 95%
2,5-Dichloropyrimidine
Purity: 95%
CAS No.: 22536-67-0
Molecular Formula: C4H2Cl2N2
Formula Weight: 148.97
Flash Point: 107°(225°F)
Physical Form: Powder
Appearance: Off-white
Melting Point: 48-55°
Beilstein Reference: 110912
UN No.: UN3077
MDL No.: MFCD00673131
Vzorec:
C₄H₂Cl₂N₂ MW: 148,97 g/mol Bod tání: 48…55 °C Bod vzplanutí: 107 °C (225 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00673131 CAS číslo: 22536-67-0 ADR: 9,III |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...