9-Phenylcarbazole-3-boronic acid 98%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
Synonyms:
(9-Phenyl-9H-carbazol-3-yl)boronic acid, 9-Phenyl-3-carbazoleboronic acid
H64059.03
H64059.06
H64059.03EA
1730
CZK
H64059.03
H64059.06
9-Phenylcarbazole-3-boronic acid 98%
9-Phenylcarbazole-3-boronic acid
Purity: 98%
CAS No.: 854952-58-2
Molecular Formula: C18H14BNO2
Formula Weight: 287.13
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Beilstein Reference: 10566519
MDL No.: MFCD12196936
Vzorec:
C₁₈H₁₄BNO₂ |
MDL Number:
MFCD12196936 CAS číslo: 854952-58-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...