trans-3-Chloro-β-nitrostyrene 97%
Dodavatel: Thermo Scientific
|
Nebezpečí
|
H64053.03
H64053.06
H64053.03EA
1180
CZK
H64053.03
H64053.06
trans-3-Chloro-β-nitrostyrene 97%
trans-3-Chloro-β-nitrostyrene
Purity: 97%
CAS No.: 37888-03-2
Molecular Formula: C8H6ClNO2
Formula Weight: 183.59
Physical Form: Powder
Appearance: Pale yellow
Melting Point: 48-52°
Beilstein Reference: 2087804
MDL No.: MFCD00175518
Vzorec:
C₈H₆ClNO₂ MW: 183,59 g/mol Bod tání: 48…52 °C Storage Temperature: Ambient |
MDL Number:
MFCD00175518 CAS číslo: 37888-03-2 |
Learn more
![](/m-cz.vwr.com/cs_CZ/images/about_vwr_275x315.jpg)
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...