6,7-Dimethoxy-3,4-dihydroisoquinoline 97%
Dodavatel: Thermo Fisher Scientific
|
Varování
|
H64018.03
H64018.06
H64018.03EA
3040
CZK
H64018.03
H64018.06
6,7-Dimethoxy-3,4-dihydroisoquinoline 97%
6,7-Dimethoxy-3,4-dihydroisoquinoline
Purity: 97%
CAS No.: 3382-18-1
Molecular Formula: C11H13NO2
Formula Weight: 191.23
Physical Form: Low melting solid
Refractive Index: 1.5930
Appearance: Yellow
Melting Point: 39-40°
Boiling Point: 155-161°/1mm
Beilstein Reference: 138269
MDL No.: MFCD00266748
Vzorec:
C₁₁H₁₃NO₂ MW: 191.23 g/mol Bod varu: 155…161 °C (1 mmHg) Bod tání: 39…40 °C Storage Temperature: Ambient |
MDL Number:
MFCD00266748 CAS číslo: 3382-18-1 EINECS: 222-185-9 |