(1-Methyl-1H-imidazol-2-yl)methanol 98%
Dodavatel: Thermo Scientific
|
Varování
|
Synonyms:
1-Methyl-2-imidazolemethanol
H64006.06
H64006.14
H64006.06EA
2150
CZK
H64006.06
H64006.14
(1-Methyl-1H-imidazol-2-yl)methanol 98%
(1-Methyl-1H-imidazol-2-yl)methanol
Purity: 98%
CAS No.: 17334-08-6
Molecular Formula: C5H8N2O
Formula Weight: 112.13
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Melting Point: 107-111°
Beilstein Reference: 113492
MDL No.: MFCD00964673
Vzorec:
C₅H₈N₂O MW: 112,13 g/mol Bod tání: 107…111 °C |
MDL Number:
MFCD00964673 CAS číslo: 17334-08-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...