Z-Hyp-OMe 98%
Dodavatel: Thermo Scientific
Synonyms:
N-Cbz-4-trans-hydroxy-L-proline methyll ester, N-Benzyloxycarbonyl-4-trans-hydroxy-L-proline methyll ester, Z-Hyp-OMe, N-Carbobenzoxy-4-trans-hydroxy-L-proline methyll ester, (2S,4R)-1-Benzyl 2-methyl 4-hydroxypyrrolidine-1,2-dicarboxylate, N-Benzyloxycarbonyl-4-trans-hydroxy-L-proline methyl ester
H63993.06
H63993.06EA
5700
CZK
H63993.06
Z-Hyp-OMe 98%
Z-Hyp-OMe
Purity: 98%
CAS No.: 64187-48-0
Molecular Formula: C14H17NO5
Formula Weight: 279.29
Storage Temperature: 2°C to 8°C
Physical Form: Liquid
Appearance: Pale yellow
MDL No.: MFCD00055851
Vzorec:
C₁₄H₁₇NO₅ MW: 279,292 g/mol Storage Temperature: Refrigerator |
MDL Number:
MFCD00055851 CAS číslo: 64187-48-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...