Tisk…

Boc-D-ABU-OH 95%

Dodavatel: Thermo Fisher Scientific

Synonyms: (R)-2-(Boc-amino)butyric acid

H63991.03 H63991.06 H63991.14
H63991.03EA 1700 CZK
H63991.03 H63991.06 H63991.14
Boc-D-ABU-OH 95%
Boc-D-ABU-OH

Purity: 95% CAS No.: 45121-22-0 Molecular Formula: C9H17NO4 Formula Weight: 203.24 Storage Temperature: -30°C to -10°C Physical Form: Liquid Appearance: Yellow MDL No.: MFCD00270335

Vzorec: C₉H₁₇NO₄
MW: 203.238 g/mol
Storage Temperature: Freezer
MDL Number: MFCD00270335
CAS číslo: 45121-22-0

Objednat


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Více About VWR